siRNA Details
Gene Symbol: LOC101927476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110386.1 | 3 | 1387 | |||
GenBank mRNA | AK123782.1 | - | 3 | 1387 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110386.1 | 3 | 1387 | |||
GenBank mRNA | AK123782.1 | - | 3 | 1387 |