siRNA Details
| Gene Symbol: GAS1RR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_049794.1 | 7 | 2647 | |||
| GenBank mRNA | BC036850.1 | - | 7 | 2656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAS1RR| Gene Symbol: GAS1RR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_049794.1 | 7 | 2647 | |||
| GenBank mRNA | BC036850.1 | - | 7 | 2656 | ||