siRNA Details
| Gene Symbol: LINC01514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120620.1 | 3 | 530 | |||
| GenBank mRNA | BC038784.1 | - | 3 | 529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01514| Gene Symbol: LINC01514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120620.1 | 3 | 530 | |||
| GenBank mRNA | BC038784.1 | - | 3 | 529 | ||