siRNA Details
Gene Symbol: LOC100506725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108082.1 | 3 | 952 | |||
NR_108083.1 | 2 | 854 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF454968.1 | - | 2 | 700 | Chr. 7 - 35716592 - 35719523 [+] on Build GRCh38 |
