siRNA Details
| Gene Symbol: COL5A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CR748640.1 | - | 2 | 231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

COL5A1-AS1| Gene Symbol: COL5A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CR748640.1 | - | 2 | 231 | ||