siRNA Details
| Gene Symbol: SPATA42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034168.1 | 2 | 327 | |||
| NR_049777.1 | 2 | 327 | ||||
| GenBank mRNA | AA401510.1 | - | 2 | 300 | ||
| AA401900.1 | - | 2 | 327 | |||
| AY312144.1 | - | 2 | 327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA42| Gene Symbol: SPATA42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034168.1 | 2 | 327 | |||
| NR_049777.1 | 2 | 327 | ||||
| GenBank mRNA | AA401510.1 | - | 2 | 300 | ||
| AA401900.1 | - | 2 | 327 | |||
| AY312144.1 | - | 2 | 327 | |||