siRNA Details
| Gene Symbol: LOC100506286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135594.1 | 2 | 1370 | |||
| GenBank mRNA | AK056547.1 | - | 2 | 1369 | ||
| BC014493.1 | - | 2 | 1348 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506286| Gene Symbol: LOC100506286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135594.1 | 2 | 1370 | |||
| GenBank mRNA | AK056547.1 | - | 2 | 1369 | ||
| BC014493.1 | - | 2 | 1348 | |||