siRNA Details
Gene Symbol: LINC01254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110775.1 | 1 | 614 | |||
GenBank mRNA | AY726567.1 | - | 1 | 614 | ||
BC040323.1 | - | 1 | 759 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110775.1 | 1 | 614 | |||
GenBank mRNA | AY726567.1 | - | 1 | 614 | ||
BC040323.1 | - | 1 | 759 |