siRNA Details
| Gene Symbol: LINC00844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108046.1 | 2 | 341 | |||
| GenBank mRNA | CB157564.1 | - | 2 | 225 | ||
| DA153602.1 | - | 2 | 258 | |||
| DB500612.1 | - | 2 | 342 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00844| Gene Symbol: LINC00844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108046.1 | 2 | 341 | |||
| GenBank mRNA | CB157564.1 | - | 2 | 225 | ||
| DA153602.1 | - | 2 | 258 | |||
| DB500612.1 | - | 2 | 342 | |||