siRNA Details
Gene Symbol: LINC00844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108046.1 | 2 | 409 | |||
GenBank mRNA | CB157564.1 | - | 2 | 293 | ||
DA153602.1 | - | 2 | 326 | |||
DB500612.1 | - | 2 | 410 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108046.1 | 2 | 409 | |||
GenBank mRNA | CB157564.1 | - | 2 | 293 | ||
DA153602.1 | - | 2 | 326 | |||
DB500612.1 | - | 2 | 410 |