siRNA Details
| Gene Symbol: LOC286059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038873.1 | 3 | 2532 | |||
| GenBank mRNA | AK024295.1 | - | 3 | 2553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC286059| Gene Symbol: LOC286059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038873.1 | 3 | 2532 | |||
| GenBank mRNA | AK024295.1 | - | 3 | 2553 | ||