siRNA Details
| Gene Symbol: LINC00301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026946.1 | 6 | 583 | |||
| GenBank mRNA | AK058123.1 | - | 6 | 583 | ||
| BC029583.1 | - | 5 | 466 | |||
| BX103095.1 | - | 5 | 469 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00301| Gene Symbol: LINC00301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026946.1 | 6 | 583 | |||
| GenBank mRNA | AK058123.1 | - | 6 | 583 | ||
| BC029583.1 | - | 5 | 466 | |||
| BX103095.1 | - | 5 | 469 | |||