siRNA Details
| Gene Symbol: LINC01563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110895.1 | 3 | 710 | |||
| NR_110896.1 | 2 | 586 | ||||
| GenBank mRNA | AB593144.1 | - | 3 | 675 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01563| Gene Symbol: LINC01563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110895.1 | 3 | 710 | |||
| NR_110896.1 | 2 | 586 | ||||
| GenBank mRNA | AB593144.1 | - | 3 | 675 | ||