siRNA Details
| Gene Symbol: RAB30-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038903.1 | 2 | 197 | |||
| GenBank mRNA | BC031316.2 | - | 2 | 187 | ||
| BU664621.1 | - | 2 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB30-AS1| Gene Symbol: RAB30-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038903.1 | 2 | 197 | |||
| GenBank mRNA | BC031316.2 | - | 2 | 187 | ||
| BU664621.1 | - | 2 | 207 | |||