siRNA Details
Gene Symbol: LINC01425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109958.1 | 4, 5 | 517 | |||
GenBank mRNA | BC035642.1 | - | 4, 5 | 527 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109958.1 | 4, 5 | 517 | |||
GenBank mRNA | BC035642.1 | - | 4, 5 | 527 |