siRNA Details
| Gene Symbol: SNHG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092096.1 | - | 2 | 1717 | ||
| BC045792.1 | - | 4 | 907 | |||
| BC064589.1 | - | 3 | 1438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNHG12| Gene Symbol: SNHG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092096.1 | - | 2 | 1717 | ||
| BC045792.1 | - | 4 | 907 | |||
| BC064589.1 | - | 3 | 1438 | |||