siRNA Details
| Gene Symbol: LOC401312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122075.1 | 3 | 1053 | |||
| GenBank mRNA | BC042871.1 | - | 3 | 1053 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC401312| Gene Symbol: LOC401312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122075.1 | 3 | 1053 | |||
| GenBank mRNA | BC042871.1 | - | 3 | 1053 | ||