siRNA Details
| Gene Symbol: LOC101928525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121579.1 | 1, 2 | 606 | |||
| GenBank mRNA | BC015688.1 | - | 1, 2 | 606 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928525| Gene Symbol: LOC101928525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121579.1 | 1, 2 | 606 | |||
| GenBank mRNA | BC015688.1 | - | 1, 2 | 606 | ||