siRNA Details
| Gene Symbol: IQCF5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109984.1 | 1 | 144 | |||
| GenBank mRNA | AK097471.1 | - | 1 | 144 | ||
| Gene Symbol: IQCF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW138546.1 | - | 1 | 35 | ||

IQCF5-AS1
Genomic Map