siRNA Details
| Gene Symbol: LINC01095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038331.1 | 4, 5 | 606 | |||
| GenBank mRNA | AK057233.1 | - | 3, 4 | 1141 | ||
| BC033400.1 | - | 4, 5 | 599 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI828405.1 | - | 3, 4 | 461 | Chr. 4 - 146111385 - 146121883 [-] on Build GRCh38 | |

LINC01095
Genomic Map