siRNA Details
| Gene Symbol: LOC101929412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104606.1 | 1 | 1507 | |||
| GenBank mRNA | AK127412.1 | - | 1 | 1507 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929412| Gene Symbol: LOC101929412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104606.1 | 1 | 1507 | |||
| GenBank mRNA | AK127412.1 | - | 1 | 1507 | ||