siRNA Details
| Gene Symbol: LINC00458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108062.1 | 4 | 915 | |||
| GenBank mRNA | BC018008.1 | - | 4 | 910 | ||
| BC020935.1 | - | 4 | 906 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00458| Gene Symbol: LINC00458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108062.1 | 4 | 915 | |||
| GenBank mRNA | BC018008.1 | - | 4 | 910 | ||
| BC020935.1 | - | 4 | 906 | |||