siRNA Details
| Gene Symbol: LINC01510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120506.1 | 3 | 546 | |||
| GenBank mRNA | CR745839.1 | - | 2 | 296 | ||
| DR422469.1 | - | 3 | 546 | |||
| LN812954.1 | - | 3 | 511 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01510| Gene Symbol: LINC01510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120506.1 | 3 | 546 | |||
| GenBank mRNA | CR745839.1 | - | 2 | 296 | ||
| DR422469.1 | - | 3 | 546 | |||
| LN812954.1 | - | 3 | 511 | |||