siRNA Details
| Gene Symbol: LOC101929294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125808.1 | 4 | 517 | |||
| GenBank mRNA | CD705133.1 | - | 2 | 194 | ||
| DA221074.1 | - | 4 | 517 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929294| Gene Symbol: LOC101929294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125808.1 | 4 | 517 | |||
| GenBank mRNA | CD705133.1 | - | 2 | 194 | ||
| DA221074.1 | - | 4 | 517 | |||