siRNA Details
| Gene Symbol: MAMDC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040076.1 | 4 | 1506 | |||
| GenBank mRNA | BC043510.1 | - | 4 | 1450 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAMDC2-AS1| Gene Symbol: MAMDC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040076.1 | 4 | 1506 | |||
| GenBank mRNA | BC043510.1 | - | 4 | 1450 | ||