siRNA Details
| Gene Symbol: CHADL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005261370.3 | XP_005261427.1 | 7 | 3050 | ||
| XM_011529932.2 | XP_011528234.1 | 6 | 3668 | |||
| Gene Symbol: LOC100506544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057177.1 | - | 3 | 1845 | ||

CHADL
Genomic Map