siRNA Details
| Gene Symbol: LOC105375304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136270.1 | 2 | 287 | |||
| NR_136271.1 | 2 | 199 | ||||
| NR_136272.1 | 2 | 94 | ||||
| GenBank mRNA | AW403608.1 | - | 2 | 206 | ||
| BE218132.1 | - | 2 | 288 | |||
| CK824920.1 | - | 2 | 152 | |||
| DB497744.1 | - | 2 | 280 | |||

LOC105375304
Genomic Map