siRNA Details
| Gene Symbol: ASB16-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_049729.1 | 4 | 2175 | |||
| NR_049730.1 | 3 | 837 | ||||
| GenBank mRNA | AK055685.1 | - | 4 | 2126 | ||
| BC009448.1 | - | 1 | 1255 | 3530891 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASB16-AS1| Gene Symbol: ASB16-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_049729.1 | 4 | 2175 | |||
| NR_049730.1 | 3 | 837 | ||||
| GenBank mRNA | AK055685.1 | - | 4 | 2126 | ||
| BC009448.1 | - | 1 | 1255 | 3530891 | ||