siRNA Details
| Gene Symbol: LINC01605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121621.1 | 3 | 404 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX104160.1 | - | 2 | 390 | Chr. 8 - 37516450 - 37517021 [-] on Build GRCh38 | |

LINC01605
Genomic Map