siRNA Details
| Gene Symbol: MIR4435-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024373.2 | 2 | 538 | |||
| GenBank mRNA | BC014776.1 | - | 2 | 465 | ||
| BC070201.1 | - | 2 | 425 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR4435-2HG| Gene Symbol: MIR4435-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024373.2 | 2 | 538 | |||
| GenBank mRNA | BC014776.1 | - | 2 | 465 | ||
| BC070201.1 | - | 2 | 425 | |||