siRNA Details
| Gene Symbol: MIR4435-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015395.2 | 7 | 3292 | |||
| NR_136161.1 | 6 | 1043 | ||||
| NR_136165.1 | 4 | 842 | ||||
| NR_136166.1 | 5 | 2875 | ||||
| GenBank mRNA | BX647931.1 | - | 6 | 931 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR4435-2HG| Gene Symbol: MIR4435-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015395.2 | 7 | 3292 | |||
| NR_136161.1 | 6 | 1043 | ||||
| NR_136165.1 | 4 | 842 | ||||
| NR_136166.1 | 5 | 2875 | ||||
| GenBank mRNA | BX647931.1 | - | 6 | 931 | ||