siRNA Details
| Gene Symbol: LOC102724927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120311.1 | 2 | 308 | |||
| GenBank mRNA | BC012429.1 | - | 2 | 308 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA435098.1 | - | 1 | 468 | Chr. 16 - 3948523 - 3949158 [-] on Build GRCh38 | |

LOC102724927
Genomic Map