siRNA Details
| Gene Symbol: DNAJB8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037890.1 | 3 | 958 | |||
| GenBank mRNA | AK097457.1 | - | 3 | 959 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJB8-AS1| Gene Symbol: DNAJB8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037890.1 | 3 | 958 | |||
| GenBank mRNA | AK097457.1 | - | 3 | 959 | ||