siRNA Details
Gene Symbol: LOC101927692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120628.1 | 3 | 472 | |||
GenBank mRNA | AK098198.1 | - | 3 | 472 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120628.1 | 3 | 472 | |||
GenBank mRNA | AK098198.1 | - | 3 | 472 |