siRNA Details
Gene Symbol: LOC105369635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135032.1 | 2 | 134 | |||
GenBank mRNA | BX094730.1 | - | 2 | 133 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM855863.1 | - | 2 | 116 | Chr. 12 - 6963264 - 6964282 [+] on Build GRCh38 |
