siRNA Details
| Gene Symbol: LOC283352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104172.1 | 4 | 401 | |||
| NR_104173.1 | 3 | 305 | ||||
| GenBank mRNA | BC035922.1 | - | 3 | 360 | ||
| BC046203.1 | - | 3 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283352| Gene Symbol: LOC283352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104172.1 | 4 | 401 | |||
| NR_104173.1 | 3 | 305 | ||||
| GenBank mRNA | BC035922.1 | - | 3 | 360 | ||
| BC046203.1 | - | 3 | 301 | |||