siRNA Details
| Gene Symbol: LOC101929634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135293.1 | 2 | 3201 | |||
| GenBank mRNA | AK128797.1 | - | 2 | 3198 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929634| Gene Symbol: LOC101929634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135293.1 | 2 | 3201 | |||
| GenBank mRNA | AK128797.1 | - | 2 | 3198 | ||