siRNA Details
| Gene Symbol: LOC105369691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135030.1 | 1 | 304 | |||
| NR_135031.1 | 1 | 304 | ||||
| GenBank mRNA | HY231162.1 | - | 1 | 343 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU570742.1 | - | 1 | 276 | Chr. 12 - 22609227 - 22625015 [-] on Build GRCh38 | |

LOC105369691
Genomic Map