siRNA Details
Gene Symbol: LOC105369691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135030.1 | 1 | 188 | |||
NR_135031.1 | 1 | 188 | ||||
GenBank mRNA | HY231162.1 | - | 1 | 227 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU570742.1 | - | 1 | 160 | Chr. 12 - 22609227 - 22625015 [-] on Build GRCh38 |
