siRNA Details
Gene Symbol: LOC100507487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125882.1 | 4 | 781 | |||
GenBank mRNA | AK093416.1 | - | 4 | 727 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100507487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125882.1 | 4 | 781 | |||
GenBank mRNA | AK093416.1 | - | 4 | 727 |