siRNA Details
Gene Symbol: LINC00708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108058.1 | 4, 5 | 581 | |||
GenBank mRNA | BC031880.1 | - | 4, 5 | 592 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG722333.1 | - | 4, 5 | 590 | Chr. 10 - 8259567 - 8268307 [-] on Build GRCh38 |
