siRNA Details
| Gene Symbol: LINC00708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108058.1 | 5 | 630 | |||
| GenBank mRNA | BC031880.1 | - | 5 | 641 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG722333.1 | - | 5 | 640 | Chr. 10 - 8259567 - 8268307 [-] on Build GRCh38 | |

LINC00708
Genomic Map