siRNA Details
Gene Symbol: LOC115110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037844.2 | 5 | 2435 | |||
GenBank mRNA | AL359943.1 | - | 1 | 1488 | ||
CR749717.1 | - | 1 | 1488 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC115110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037844.2 | 5 | 2435 | |||
GenBank mRNA | AL359943.1 | - | 1 | 1488 | ||
CR749717.1 | - | 1 | 1488 |