siRNA Details
Gene Symbol: LINC00485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033855.1 | 5 | 2741 | |||
GenBank mRNA | BC037211.1 | - | 5 | 2740 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033855.1 | 5 | 2741 | |||
GenBank mRNA | BC037211.1 | - | 5 | 2740 |