siRNA Details
Gene Symbol: KCNMB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126560.1 | 4 | 357 | |||
NR_126561.1 | 4 | 445 | ||||
GenBank mRNA | AF279780.1 | - | 4 | 440 | ||
AY769439.1 | - | 4 | 304 | |||
BG206373.1 | - | 2 | 201 | |||
DA735220.1 | - | 6 | 518 | |||
U61097.1 | - | 3 | 251 |
