siRNA Details
Gene Symbol: LOC101927132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110650.1 | 6 | 2383 | |||
GenBank mRNA | AK091370.1 | - | 6 | 2385 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110650.1 | 6 | 2383 | |||
GenBank mRNA | AK091370.1 | - | 6 | 2385 |