siRNA Details
Gene Symbol: LOC100505797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134647.1 | 2, 3 | 258 | |||
GenBank mRNA | BX114234.1 | - | 2, 3 | 258 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG217098.1 | - | 1, 2 | 162 | Chr. 18 - 72880103 - 72881399 [+] on Build GRCh38 |
