siRNA Details
| Gene Symbol: HMGN3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040671.1 | 2 | 1249 | |||
| GenBank mRNA | BC042096.1 | - | 2 | 1206 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK095853.1 | - | 2 | 1333 | Chr. 6 - 79233700 - 79236080 [+] on Build GRCh38 | |

HMGN3-AS1
Genomic Map