siRNA Details
Gene Symbol: MIR3663HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121650.1 | 2 | 245 | |||
NR_121651.1 | 2 | 245 | ||||
GenBank mRNA | BC039338.1 | - | 2 | 245 | ||
HY017741.1 | - | 2 | 238 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIR3663HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121650.1 | 2 | 245 | |||
NR_121651.1 | 2 | 245 | ||||
GenBank mRNA | BC039338.1 | - | 2 | 245 | ||
HY017741.1 | - | 2 | 238 |