siRNA Details
| Gene Symbol: LOC101929450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125814.1 | 4 | 712 | |||
| NR_125815.1 | 4 | 1211 | ||||
| GenBank mRNA | BC015784.1 | - | 4 | 1166 | ||
| BC047326.1 | - | 4 | 1170 | |||
| BC082237.1 | - | 4 | 696 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929450| Gene Symbol: LOC101929450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125814.1 | 4 | 712 | |||
| NR_125815.1 | 4 | 1211 | ||||
| GenBank mRNA | BC015784.1 | - | 4 | 1166 | ||
| BC047326.1 | - | 4 | 1170 | |||
| BC082237.1 | - | 4 | 696 | |||